Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thoc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK040311.1 | - | 1 | 603 | ||
AK087099.1 | - | 1 | 583 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB664125.1 | - | 1 | 583 | Chr. 11 - 4895360 - 4895998 [+] on Build GRCm38 |