Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmppe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK082033.1 | - | 1 | 650 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU694508.1 | - | 1 | 489 | Chr. 9 - 114409812 - 114410345 [+] on Build GRCm38 |