Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930579J19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016326.1 | - | 1 | 185 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715965.1 | - | 185 | Chr. - - [-] on Build |