Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4833447P13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019531.1 | - | 2317 | |||
AK042445.1 | - | 1930 | ||||
BB242542.2 | - | 250 |
Gene Symbol: Slc12a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK085496.1 | - | 24 | 3985 |