Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gid8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA726152.1 | - | 1 | 324 | ||
AK076477.1 | - | 1 | 865 | |||
AK143107.1 | - | 1 | 867 | |||
AV250006.2 | - | 1 | 39 |