Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gid8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076477.1 | - | 1 | 1359 | ||
AK143107.1 | - | 1 | 1361 | |||
AV250006.2 | - | 1 | 533 |