Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK089397.1 | - | 1 | 1503 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY764398.1 | - | 1 | 654 | Chr. 17 - 15500768 - 15501482 [+] on Build GRCm38 |