Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK042883.1 | - | 78 | |||
AK054023.1 | - | 68 |
Gene Symbol: LOC102639683 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK085371.1 | - | 78 |