Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sap130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK149280.1 | - | 342 | |||
AK172277.1 | - | 2 | 738 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY742164.1 | - | 2 | 342 | Chr. 18 - 31635101 - 31636611 [+] on Build GRCm38 |