Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1cl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC028632.1 | - | 3 | 846 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK054287.1 | - | 482 | Chr. - - [-] on Build |