Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4632404H12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK040741.1 | - | 1 | 1673 | ||
AK040882.1 | - | 1 | 1701 | |||
AK047674.1 | - | 1 | 1690 | |||
AK048101.1 | - | 1 | 1859 | |||
AK084338.1 | - | 1 | 1691 | |||
AK164170.1 | - | 1 | 1750 | |||
AK169469.1 | - | 1 | 1698 | |||
BC063049.1 | - | 1 | 1250 | |||
BC080683.1 | - | 1 | 1689 |