Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srcap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK081131.1 | - | 1289 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB352865.2 | - | 1 | 231 | Chr. 7 - 127566515 - 127567054 [+] on Build GRCm38 |