Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srcap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK081131.1 | - | 674 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG806293.1 | - | 1 | 579 | Chr. 7 - 127565552 - 127566151 [+] on Build GRCm38 |