Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Copg2os2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AF217545.1 | - | LIT1225 Mit1/Lb9 |
1 | 1659 | |
AK014219.1 | - | 1 | 20 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AF177020.1 | - | 1 | 307 | Chr. - - [-] on Build |