Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005616.1 | - | 1 | 365 | ||
AK079015.1 | - | 1 | 376 | |||
AK225018.1 | - | 1 | 366 |