Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm16532 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029648.1 | - | 238 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714745.1 | - | 2 | 235 | Chr. 9 - 21152538 - 21166880 [-] on Build GRCm38 |