Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173520.2 | NP_775791.1 | 2 | 536 | ||
GenBank mRNA | BC034752.1 | - | 2 | 536 | 5193354 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000623103.1 | - | 536 | Chr. - - [-] on Build |