Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHG4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006714446.3 | XP_006714509.1 | 2 | 205 | ||
XM_011513968.2 | XP_011512270.1 | 2 | 205 | |||
XM_011513969.2 | XP_011512271.1 | 2 | 205 | |||
XM_011513971.2 | XP_011512273.1 | 2 | 150 | |||
XM_011513972.2 | XP_011512274.1 | 2 | 206 | |||
XM_011513973.1 | XP_011512275.1 | 2 | 203 | |||
XM_017009112.1 | XP_016864601.1 | 2 | 206 |