Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MICAL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126925.1 | - | 1 | 330 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000476405.1 | - | 1 | 330 | Chr. 22 - 17787652 - 17811497 [-] on Build GRCh38 |