Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGAP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB075855.1 | - | 4 | 1544 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000444431.1 | - | 4 | 1544 | Chr. 10 - 86992406 - 87010203 [+] on Build GRCh38 |