Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C10orf25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039380.3 | NP_001034469.2 | 2 | 1408 | ||
NR_126421.1 | 3 | 545 | ||||
XM_017015895.1 | XP_016871384.1 | 3 | 539 | |||
GenBank mRNA | AK055129.1 | - | 2 | 1386 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000625168.1 | - | 1386 | Chr. - - [-] on Build |