Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHANK2-AS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073536.1 | 3 | 1148 | |||
GenBank mRNA | BC004224.2 | - | 3 | 1148 | 3537945 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000307548.2 | - | 2 | 1148 | Chr. 11 - 70863283 - 70865114 [+] on Build GRCh38 |