Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR4435-2HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015395.2 | 7 | 3756 | |||
NR_136161.1 | 6 | 1507 | ||||
NR_136165.1 | 4 | 1306 | ||||
NR_136166.1 | 5 | 3339 | ||||
GenBank mRNA | BX647931.1 | - | 6 | 1395 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000439362.5 | - | 6 | 1394 | Chr. 2 - 111207776 - 111495048 [-] on Build GRCh38 |