Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026939.1 | 1, 2 | 288 | |||
NR_026940.1 | 1, 2 | 288 | ||||
NR_026941.1 | 1, 2 | 288 | ||||
GenBank mRNA | AJ344228.1 | - | 1, 2 | 101 | ||
AJ535620.1 | - | 1, 2 | 101 | |||
AJ535621.1 | - | 1, 2 | 101 | |||
AJ535622.1 | - | 1, 2 | 115 | |||
AK055184.1 | - | 1, 2 | 115 | |||
AY226450.1 | - | 1, 2 | 104 | |||
AY226451.1 | - | 1, 2 | 104 | |||
BC036393.1 | - | 1, 2 | 290 | |||
BC112306.1 | - | 12 | ||||
BC112308.1 | - | 12 | ||||
BC112326.1 | - | 1, 2 | 108 |