Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7orf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026865.1 | 1 | 1163 | |||
GenBank mRNA | AF063598.1 | - | 1 | 930 | ||
AF439977.1 | - | 1 | 1163 | |||
AK056371.1 | - | 1 | 1079 | |||
BC031037.1 | - | 1 | 1048 | |||
BC070228.1 | - | 2 | 488 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000435354.1 | - | 2 | 485 | Chr. 7 - 156639432 - 156640563 [-] on Build GRCh38 |