Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2orf27B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_214461.2 | NP_999626.1 | 2 | 416 | ||
GenBank mRNA | BC043584.1 | - | 2 | 416 | 5229449 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000623129.1 | - | 416 | Chr. - - [-] on Build |