Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSAP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY176665.1 | - | 3 | 527 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000524252.1 | - | 3 | 527 | Chr. 8 - 18720905 - 18734405 [+] on Build GRCh38 |