Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTBP1-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK056133.1 | - | 1 | 665 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000578730.1 | - | 1 | 646 | Chr. 4 - 1249451 - 1288291 [+] on Build GRCh38 |