Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSORS1C3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026816.1 | 1 | 91 | |||
GenBank mRNA | AB932952.1 | - | 1 | 91 | ||
AB932953.1 | - | 1 | 91 | |||
AB932954.1 | - | 1 | 91 | |||
AB933312.1 | - | 1 | 91 | |||
AB933313.1 | - | 1 | 91 | |||
AB933320.1 | - | 1 | 91 | |||
AY484516.1 | - | 1 | 91 | |||
LC027944.1 | - | 1 | 91 | |||
LC027945.1 | - | 2 | 383 | |||
LC027946.1 | - | 2 | 383 | |||
LC050979.1 | - | 2 | 383 | |||
LC050980.1 | - | 2 | 383 | |||
LC050981.1 | - | 2 | 383 | |||
LC050982.1 | - | 2 | 383 | |||
LC050983.1 | - | 2 | 383 | |||
LC050985.1 | - | 2 | 383 | |||
LC050986.1 | - | 2 | 383 | |||
LC050987.1 | - | 2 | 383 | |||
LC050989.1 | - | 2 | 383 | |||
LC050990.1 | - | 2 | 383 | |||
LC050991.1 | - | 2 | 383 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000412143.1 | - | 1 | 91 | Chr. 6 - 31173735 - 31177899 [-] on Build GRCh38 |