Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1QTNF9B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014442.2 | NP_001014442.2 | 3, 4 | 311 | ||
GenBank mRNA | BC015452.1 | - | 3, 4 | 313 | ||
BC073902.1 | - | 3, 4 | 311 | |||
BC132842.1 | - | 1, 2 | 147 | |||
BC132846.1 | - | 1, 2 | 147 |