Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040662.1 | 2 | 943 | |||
GenBank mRNA | AK093953.1 | - | 4 | 828 | ||
AK290875.1 | - | 2 | 938 | |||
BC106759.2 | - | 2 | 889 | 40024782 | ||
BC113853.1 | - | 1 | 980 | |||
BC114462.1 | - | 2 | 889 | |||
BX640979.1 | - | 2 | 941 | |||
L06175.1 | - | 2 | 934 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000541196.2 | - | 2 | 941 | Chr. 6 - 31463185 - 31464379 [+] on Build GRCh38 |