Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EGFR-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047551.1 | 2 | 2193 | |||
GenBank mRNA | AK123474.1 | - | 2 | 2178 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000442411.1 | - | 2 | 2178 | Chr. 7 - 55179750 - 55188934 [-] on Build GRCh38 |