Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105374266 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135537.1 | 2 | 1419 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000446091.1 | - | 2 | 1419 | Chr. 3 - 187965768 - 187976407 [-] on Build GRCh38 |