Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC440446 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122104.1 | 3 | 1504 | |||
GenBank mRNA | AK124832.1 | - | 3 | 1504 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000441895.2 | - | 3 | 1504 | Chr. 17 - 51336715 - 51342571 [+] on Build GRCh38 |