Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM66C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026788.1 | 5 | 1584 | |||
GenBank mRNA | AK096301.1 | - | 4 | 1448 | ||
AK311411.1 | - | 3 | 801 | |||
BC034777.1 | - | 5 | 1524 | |||
BC048994.1 | - | 3 | 941 |