Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOX21-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046514.1 | 2 | 1613 | |||
GenBank mRNA | AK055459.1 | - | 2 | 1615 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000438290.2 | - | 2 | 1613 | Chr. 13 - 94712716 - 94716246 [+] on Build GRCh38 |