Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00837 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038374.1 | 3 | 786 | |||
NR_038375.1 | 2 | 684 | ||||
NR_038376.1 | 2 | 563 | ||||
GenBank mRNA | BC028422.1 | - | 2 | 608 | ||
DB099527.1 | - | 2 | 520 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000443246.2 | - | 2 | 640 | Chr. 10 - 28789188 - 28796006 [-] on Build GRCh38 |