Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKN2B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003529.3 | 19 | 3296 | |||
NR_047532.1 | 14 | 2085 | ||||
NR_047534.1 | 9 | 1349 | ||||
NR_047535.1 | 8 | 1244 | ||||
NR_047536.1 | 7 | 1108 | ||||
NR_047537.1 | 6 | 1044 | ||||
NR_047538.1 | 5 | 908 | ||||
NR_047543.1 | 9 | 1378 | ||||
NR_120536.1 | 4 | 697 | ||||
GenBank mRNA | AB548314.1 | - | 14 | 2085 | ||
BC038540.1 | - | 3 | 386 | |||
DQ485453.1 | - | 19 | 3295 | |||
GQ495918.1 | - | 4 | 586 | |||
GQ495919.1 | - | 5 | 797 | |||
GQ495920.1 | - | 7 | 997 | |||
GQ495921.1 | - | 6 | 933 | |||
GQ495922.1 | - | 8 | 1133 | |||
GQ495923.1 | - | 9 | 1267 |