Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKN2B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003529.3 | 6 | 1222 | |||
NR_047532.1 | 5 | 909 | ||||
NR_047533.1 | 3 | 614 | ||||
NR_047534.1 | 3 | 614 | ||||
NR_047535.1 | 3 | 614 | ||||
NR_047536.1 | 3 | 614 | ||||
NR_047537.1 | 3 | 614 | ||||
NR_047538.1 | 3 | 614 | ||||
NR_047539.1 | 6 | 1222 | ||||
NR_047540.1 | 3 | 614 | ||||
NR_047541.1 | 3 | 614 | ||||
NR_047542.1 | 3 | 614 | ||||
NR_047543.1 | 3 | 614 | ||||
NR_120536.1 | 3 | 614 | ||||
GenBank mRNA | AB548314.1 | - | 5 | 909 | ||
DQ485453.1 | - | 6 | 1221 | |||
DQ485454.1 | - | 6 | 1221 | |||
EU741058.1 | - | 3 | 310 | |||
GQ422823.1 | - | 3 | 614 | |||
GQ495918.1 | - | 3 | 503 | |||
GQ495919.1 | - | 3 | 503 | |||
GQ495920.1 | - | 3 | 503 | |||
GQ495921.1 | - | 3 | 503 | |||
GQ495922.1 | - | 3 | 503 | |||
GQ495923.1 | - | 3 | 503 | |||
GQ495924.1 | - | 3 | 503 | |||
GQ495925.1 | - | 3 | 614 | |||
GQ495926.1 | - | 3 | 614 |