Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXP1-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK025793.1 | - | 2 | 1467 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000498714.1 | - | 2 | 1467 | Chr. 3 - 71570255 - 71574457 [-] on Build GRCh38 |