Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BDNF-AS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002832.2 | 4 | 360 | |||
NR_033312.1 | 3 | 291 | ||||
NR_033313.1 | 3 | 291 | ||||
NR_033314.1 | 4 | 360 | ||||
NR_033315.1 | 3 | 291 | ||||
GenBank mRNA | AY054391.4 | - | 4 | 390 | ||
AY054401.3 | - | 3 | 291 | |||
AY054402.3 | - | 3 | 291 | |||
AY054403.4 | - | 4 | 390 | |||
AY054404.4 | - | 3 | 291 | |||
AY054405.4 | - | 4 | 390 | |||
AY513486.3 | - | 3 | 291 | |||
EF689022.1 | - | 3 | 296 | |||
EF689023.1 | - | 3 | 296 | |||
EF689024.1 | - | 2 | 199 | |||
EF689025.1 | - | 2 | 199 | |||
EF689026.1 | - | 2 | 199 | |||
EF689027.1 | - | 2 | 199 | |||
EF689028.1 | - | 2 | 199 | |||
EF689029.1 | - | 2 | 199 | |||
EF689030.1 | - | 4 | 365 | |||
EF689031.1 | - | 4 | 365 | |||
EF689032.1 | - | 4 | 365 | |||
EF689033.1 | - | 4 | 365 | |||
EF689034.1 | - | 4 | 365 | |||
EF689035.1 | - | 4 | 365 | |||
EF689036.1 | - | 3 | 296 | |||
EF689037.1 | - | 3 | 296 | |||
EF689038.1 | - | 3 | 296 | |||
EF689039.1 | - | 3 | 296 | |||
EF689040.1 | - | 3 | 296 | |||
EF689041.1 | - | 3 | 296 | |||
EF689042.1 | - | 4 | 365 |