Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROPN1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006714503.2 | XP_006714566.1 | 6 | 1306 | ||
XM_006714504.2 | XP_006714567.1 | 7 | 1208 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000416930.2 | - | 2 | 433 | Chr. 5 - 10479371 - 10482309 [+] on Build GRCh38 |