Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100128494 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104178.1 | 1 | 2201 | |||
GenBank mRNA | AK126629.1 | - | 1 | 2201 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000502284.1 | - | 1 | 2201 | Chr. 11 - 72014291 - 72020910 [+] on Build GRCh38 |