Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MALAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002819.3 | 1 | 6650 | |||
GenBank mRNA | EF177381.1 | - | 1 | 6650 | ||
FJ209305.1 | - | 1 | 5367 |