Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXA10-AS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046609.1 | 3 | 711 | |||
GenBank mRNA | BM549446.1 | - | 3 | 683 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000523790.1 | - | 2 | 829 | Chr. 7 - 27169094 - 27171765 [+] on Build GRCh38 |