Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD21-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033886.1 | 2 | 1419 | |||
GenBank mRNA | BC031280.1 | - | 2 | 1419 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000521487.2 | - | 2 | 1419 | Chr. 8 - 116874424 - 116876868 [+] on Build GRCh38 |