Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERICH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017013135.1 | XP_016868624.1 | 1 | 559 | ||
XM_017013136.1 | XP_016868625.1 | 1 | 559 | |||
XM_017013137.1 | XP_016868626.1 | 1 | 559 | |||
XM_017013138.1 | XP_016868627.1 | 1 | 559 | |||
XM_017013139.1 | XP_016868628.1 | 1 | 559 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000520760.1 | - | 1 | 43 | Chr. 8 - 725188 - 725877 [-] on Build GRCh38 |