Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100128233 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103769.1 | 4 | 1065 | |||
GenBank mRNA | BX161428.1 | - | 4 | 1065 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000531609.1 | - | 4 | 1065 | Chr. 14 - 65212893 - 65222347 [-] on Build GRCh38 |