Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105369266 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC042588.1 | - | 1 | 665 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000566232.1 | - | 1 | 831 | Chr. 16 - 32262827 - 32265514 [-] on Build GRCh38 |