Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCCAT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027756.1 | 1 | 175 | |||
GenBank mRNA | FJ222407.1 | - | 1 | 175 | ||
FJ222408.1 | - | 1 | 175 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000569990.2 | - | 1 | 175 | Chr. 16 - 73092349 - 73093774 [+] on Build GRCh38 |